Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INSR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008249177.2 | XP_008247399.1 | 2738 | 83 | ||
XM_017337973.1 | XP_017193462.1 | 2738 | 83 | |||
XM_017337974.1 | XP_017193463.1 | 2702 | 83 | |||
XM_017337975.1 | XP_017193464.1 | 2702 | 83 | |||
GenBank mRNA | AY339877.1 | - | 2558 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus |