Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHRD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008266589.1 | XP_008264811.1 | 11 - 12 | 2258 | 102 | |
XM_017346951.1 | XP_017202440.1 | 11 - 12 | 2258 | 102 | ||
XM_017346952.1 | XP_017202441.1 | 2258 | 102 | |||
GenBank mRNA | AY575210.1 | - | 2 - 3 | 367 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |