Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718166.3 | XP_002718212.1 | 2 - 3 | 252 | 144 | |
GenBank mRNA | D82876.1 | - | 2 - 3 | 210 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |