Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718143.4 | XP_002718189.1 | 1 - 2 | 186 | 84 | |
GenBank mRNA | DQ237914.1 | - | 1 - 2 | 52 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |