Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082012.1 | NP_001075481.1 | 4 - 5 | 530 | 98 | |
XM_008263945.2 | XP_008262167.1 | 4 - 5 | 516 | 98 | ||
GenBank mRNA | AB115085.1 | - | 4 - 5 | 530 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Oryctolagus cuniculus Probe spans exons |