Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ODF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082046.1 | NP_001075515.1 | 1210 | 96 | ||
GenBank mRNA | AF107202.1 | - | 1210 | 96 | ||
AJ130873.1 | - | 1210 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Oryctolagus cuniculus |