Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LALBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082052.1 | NP_001075521.1 | 2 - 3 | 322 | 82 | |
XM_008256394.2 | XP_008254616.1 | 3 - 4 | 389 | 82 | ||
XM_017341618.1 | XP_017197107.1 | 3 - 4 | 355 | 82 | ||
GenBank mRNA | AF124257.1 | - | 2 - 3 | 322 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |