Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082054.1 | NP_001075523.1 | 8 - 9 | 1068 | 60 | |
GenBank mRNA | AF131883.1 | - | 8 - 9 | 1068 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |