Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082065.1 | NP_001075534.1 | 10 - 11 | 975 | 88 | |
GenBank mRNA | AF170528.1 | - | 10 - 11 | 975 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |