Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNMA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082070.1 | NP_001075539.1 | 17 - 18 | 2012 | 105 | |
GenBank mRNA | AF201702.1 | - | 17 - 18 | 2012 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |