Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCY10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082153.1 | NP_001075622.1 | 17 - 18 | 2473 | 136 | |
XM_008263960.2 | XP_008262182.1 | 17 - 18 | 2827 | 136 | ||
GenBank mRNA | AY212921.1 | - | 17 - 18 | 2473 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |