Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCY10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082153.1 | NP_001075622.1 | 4 - 5 | 593 | 80 | |
XM_008263960.2 | XP_008262182.1 | 4 - 5 | 947 | 80 | ||
GenBank mRNA | AY212921.1 | - | 4 - 5 | 593 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |