Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082200.1 | NP_001075669.1 | 2 - 3 | 153 | 101 | |
GenBank mRNA | D21265.1 | - | 2 - 3 | 153 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |