Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082204.1 | NP_001075673.1 | 3 - 4 | 277 | 70 | |
GenBank mRNA | D29666.1 | - | 3 - 4 | 277 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Oryctolagus cuniculus Probe spans exons |