Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KITLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082223.1 | NP_001075692.1 | 6 - 7 | 715 | 125 | |
GenBank mRNA | DQ356265.1 | - | 6 - 7 | 715 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |