Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082315.1 | NP_001075784.1 | 12 - 13 | 1565 | 77 | |
XM_008248341.2 | XP_008246563.1 | 12 - 13 | 1627 | 77 | ||
XM_008253164.2 | XP_008251386.1 | 11 - 12 | 1527 | 77 | ||
GenBank mRNA | M95815.1 | - | 12 - 13 | 1565 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |