Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC15A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082700.1 | NP_001076169.1 | 10 - 11 | 968 | 101 | |
XM_017346648.1 | XP_017202137.1 | 9 - 10 | 935 | 101 | ||
GenBank mRNA | U32507.1 | - | 10 - 11 | 968 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |