Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082703.2 | NP_001076172.1 | 11 - 12 | 1477 | 74 | |
XM_008257419.1 | XP_008255641.1 | 12 - 13 | 1550 | 74 | ||
XM_017342181.1 | XP_017197670.1 | 7 - 8 | 1737 | 74 | ||
GenBank mRNA | U55053.1 | - | 11 - 12 | 1478 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |