Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082712.1 | NP_001076181.1 | 2 - 3 | 122 | 72 | |
XM_008268542.2 | XP_008266764.1 | 2 - 3 | 183 | 72 | ||
XM_008268549.2 | XP_008266771.2 | 2 - 3 | 557 | 72 | ||
GenBank mRNA | AF040387.1 | - | 2 - 3 | 122 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Oryctolagus cuniculus Probe spans exons |