Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082642.1 | NP_001076111.1 | 9 - 10 | 1591 | 67 | |
XM_017344777.1 | XP_017200266.1 | 9 - 10 | 1659 | 67 | ||
XM_017344778.1 | XP_017200267.1 | 10 - 11 | 1394 | 67 | ||
GenBank mRNA | DQ146940.1 | - | 9 - 10 | 1591 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Oryctolagus cuniculus Probe spans exons |