Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITIH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082647.2 | NP_001076116.1 | 17 - 18 | 2320 | 131 | |
XM_008268046.2 | XP_008266268.1 | 16 - 17 | 2326 | 131 | ||
GenBank mRNA | AB050592.1 | - | 17 - 18 | 2322 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |