Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITIH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082647.2 | NP_001076116.1 | 7 - 8 | 849 | 61 | |
XM_008268046.2 | XP_008266268.1 | 6 - 7 | 855 | 61 | ||
GenBank mRNA | AB050592.1 | - | 7 - 8 | 851 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Oryctolagus cuniculus Probe spans exons |