Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082651.1 | NP_001076120.1 | 3 - 4 | 532 | 106 | |
XM_008266183.2 | XP_008264405.1 | 4 - 5 | 1114 | 106 | ||
XM_008266184.2 | XP_008264406.1 | 5 - 6 | 839 | 106 | ||
XM_017346639.1 | XP_017202128.1 | 2 - 3 | 358 | 106 | ||
GenBank mRNA | AF170493.1 | - | 3 - 4 | 532 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |