Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082756.1 | NP_001076225.1 | 10 - 11 | 1012 | 140 | |
XM_017346650.1 | XP_017202139.1 | 10 - 11 | 1017 | 140 | ||
XM_017346651.1 | XP_017202140.1 | 8 - 9 | 753 | 140 | ||
GenBank mRNA | U46915.1 | - | 10 - 11 | 1012 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Oryctolagus cuniculus Probe spans exons |