Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC10A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082764.1 | NP_001076233.1 | 2 - 3 | 499 | 71 | |
GenBank mRNA | Z54357.1 | - | 540 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Oryctolagus cuniculus Probe spans exons |