Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2CA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101686.1 | NP_001095156.1 | 1632 | 106 | ||
GenBank mRNA | X06087.1 | - | 1632 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Oryctolagus cuniculus |