Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101689.1 | NP_001095159.1 | 13 - 14 | 5016 | 87 | |
GenBank mRNA | X07995.1 | - | 13 - 14 | 5016 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |