Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPCN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143934.1 | NP_001137406.1 | 5 - 6 | 621 | 75 | |
GenBank mRNA | EU344155.1 | - | 5 - 6 | 621 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Oryctolagus cuniculus Probe spans exons |