Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082281.1 | NP_001075750.1 | 1848 | 87 | ||
XM_008249591.2 | XP_008247813.1 | 724 | 87 | |||
GenBank mRNA | J04480.1 | - | 702 | 87 | ||
M25750.1 | - | 1848 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Oryctolagus cuniculus |