Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL18R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164863.1 | NP_001158335.1 | 8 - 9 | 1108 | 95 | |
XM_008252716.2 | XP_008250938.1 | 9 - 10 | 1657 | 95 | ||
GenBank mRNA | EU731009.1 | - | 8 - 9 | 1108 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Oryctolagus cuniculus Probe spans exons |