Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199368.1 | NP_001186297.1 | 8 - 9 | 1379 | 56 | |
GenBank mRNA | FJ429175.2 | - | 8 - 9 | 1379 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Oryctolagus cuniculus Probe spans exons |