Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001171376.1 | NP_001164847.1 | 234 | 74 | ||
XM_017338558.1 | XP_017194047.1 | 588 | 74 | |||
XM_017338559.1 | XP_017194048.1 | 588 | 74 | |||
XM_017338560.1 | XP_017194049.1 | 588 | 74 | |||
XM_017338561.1 | XP_017194050.1 | 561 | 74 | |||
XM_017338562.1 | XP_017194051.1 | 586 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Oryctolagus cuniculus |