Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195764.1 | NP_001182693.1 | 1301 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Oryctolagus cuniculus |