Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENOK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199833.1 | NP_001186762.1 | 2 - 3 | 105 | 109 | |
GenBank mRNA | DN886114.1 | - | 2 - 3 | 105 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Oryctolagus cuniculus Probe spans exons |