Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SELENOK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199833.1 | NP_001186762.1 | 4 - 5 | 284 | 69 | |
GenBank mRNA | DN886114.1 | - | 4 - 5 | 284 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Oryctolagus cuniculus Probe spans exons |