Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NME2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204750.1 | NP_001191679.1 | 2 - 3 | 166 | 84 | |
GenBank mRNA | EC618915.1 | - | 2 - 3 | 174 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Oryctolagus cuniculus Probe spans exons |