Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDSN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105169.1 | NP_001098639.1 | 89 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |