Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015110694.2 | XP_014966180.1 | 341 | 67 | ||
XM_015110695.2 | XP_014966181.1 | 682 | 67 | |||
XM_028830963.1 | XP_028686796.1 | 399 | 67 | |||
XM_028830964.1 | XP_028686797.1 | 712 | 67 | |||
XM_028830966.1 | XP_028686799.1 | 703 | 67 | |||
XM_028830967.1 | XP_028686800.1 | 314 | 67 | |||
GenBank mRNA | JU332938.1 | - | 307 | 67 | ||
JV046205.1 | - | 226 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |