Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU334587.1 | - | 6688 | 89 | ||
JV715523.1 | - | 6711 | 89 | |||
JV715524.1 | - | 6717 | 89 |
Gene Symbol: CLVS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265738.1 | NP_001252667.1 | 1037 | 89 | ||
GenBank mRNA | JV564997.1 | - | 1037 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |