Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALHM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194618.2 | NP_001181547.2 | 279 | 99 | ||
XM_015148024.2 | XP_015003510.2 | 3152 | 99 | |||
XM_028853524.1 | XP_028709357.1 | 2819 | 99 | |||
XM_028853525.1 | XP_028709358.1 | 3870 | 99 | |||
XM_028853526.1 | XP_028709359.1 | 3390 | 99 | |||
XM_028853527.1 | XP_028709360.1 | 4063 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |