Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAAO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001111024.4 | XP_001111024.2 | 1189 | 72 | ||
XM_028831720.1 | XP_028687553.1 | 565 | 72 | |||
XM_028831721.1 | XP_028687554.1 | 565 | 72 | |||
XM_028831722.1 | XP_028687555.1 | 470 | 72 | |||
XM_028831723.1 | XP_028687556.1 | 464 | 72 | |||
XM_028831725.1 | XP_028687558.1 | 1143 | 72 | |||
XM_028831727.1 | XP_028687560.1 | 634 | 72 | |||
GenBank mRNA | JV678509.1 | - | 501 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |