Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAAO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001111024.4 | XP_001111024.2 | 1533 | 65 | ||
XM_028831720.1 | XP_028687553.1 | 909 | 65 | |||
XM_028831721.1 | XP_028687554.1 | 909 | 65 | |||
XM_028831722.1 | XP_028687555.1 | 814 | 65 | |||
XM_028831723.1 | XP_028687556.1 | 808 | 65 | |||
XM_028831724.1 | XP_028687557.1 | 821 | 65 | |||
XM_028831725.1 | XP_028687558.1 | 1487 | 65 | |||
XM_028831726.1 | XP_028687559.1 | 777 | 65 | |||
XM_028831727.1 | XP_028687560.1 | 978 | 65 | |||
GenBank mRNA | JU478147.1 | - | 817 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |