Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATRAID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193926.2 | NP_001180855.1 | 533 | 68 | ||
XM_028831453.1 | XP_028687286.1 | 563 | 68 | |||
XM_028831454.1 | XP_028687287.1 | 319 | 68 | |||
GenBank mRNA | JU334872.1 | - | 546 | 68 | ||
JU487868.1 | - | 365 | 68 | |||
JV043224.1 | - | 500 | 68 |
Gene Symbol: SLC5A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV664632.1 | - | 708 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |