Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PALM2AKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204121.2 | NP_001191050.1 | 315 | 73 | ||
NM_001371502.1 | NP_001358431.1 | 189 | 73 | |||
GenBank mRNA | JU332794.1 | - | 189 | 73 | ||
JU334592.1 | - | 189 | 73 |