Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193695.3 | NP_001180624.1 | 945 | 98 | ||
GenBank mRNA | JU327647.1 | - | 78 | 98 | ||
JU478647.1 | - | 149 | 98 | |||
JV705016.1 | - | 250 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |