Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201567.3 | NP_001188496.1 | 831 | 76 | ||
NM_001361229.2 | NP_001348158.1 | 856 | 76 | |||
NM_001361230.2 | NP_001348159.1 | 1069 | 76 | |||
GenBank mRNA | CN805358.1 | - | 362 | 76 | ||
JV601791.1 | - | 1069 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |