Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194410.2 | NP_001181339.1 | 310 | 84 | ||
XM_015134515.2 | XP_014990001.1 | 458 | 84 | |||
XM_015134516.2 | XP_014990002.1 | 242 | 84 | |||
XM_028845034.1 | XP_028700867.1 | 297 | 84 | |||
GenBank mRNA | JU335951.1 | - | 303 | 84 | ||
JU476832.1 | - | 281 | 84 | |||
JV047955.1 | - | 291 | 84 | |||
JV478827.1 | - | 310 | 84 | |||
JV591006.1 | - | 310 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |