Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194511.2 | NP_001181440.1 | 198 | 63 | ||
XM_001111285.4 | XP_001111285.1 | 156 | 63 | |||
GenBank mRNA | JU320131.1 | - | 160 | 63 | ||
JU320132.1 | - | 159 | 63 | |||
JU473417.1 | - | 189 | 63 | |||
JV691979.1 | - | 182 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |