Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258129.1 | NP_001245058.1 | 912 | 128 | ||
XM_015121180.2 | XP_014976666.1 | 1279 | 128 | |||
GenBank mRNA | JU327687.1 | - | 763 | 128 | ||
JU471196.1 | - | 912 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |