Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257748.1 | NP_001244677.1 | 666 | 79 | ||
XM_028834394.1 | XP_028690227.1 | 711 | 79 | |||
GenBank mRNA | JU321740.1 | - | 666 | 79 | ||
JU321741.1 | - | 666 | 79 | |||
JU473805.1 | - | 666 | 79 | |||
JV046785.1 | - | 676 | 79 | |||
JV639369.1 | - | 669 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |